Learn More
Aviva Systems Biology Corp HSF2BP Antibody - N-terminal region (ARP34838_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34838_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSF2BP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSF2BP. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55582