Learn More
Aviva Systems Biology Corp HSFX1 Antibody - middle region (ARP35906_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35906_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSFX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSFX1. Alternate names: LW-1, HSFX2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55919