Learn More
Aviva Systems Biology Corp HSFY1 Antibody - N-terminal region (ARP36095_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36095_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSFY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSFY1. Alternate names: HSFY, HSF2L, HSFY2ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-56012