Learn More
Aviva Systems Biology Corp HSFY1 Antibody - middle region (ARP39746_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39746_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSFY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HSFY1. Alternate names: HSF2L, HSFY, HSFY2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-57164