Learn More
Aviva Systems Biology Corp HSFY1 Antibody - N-terminal region (ARP39745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSFY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HSFY1. Alternate names: HSF2L, HSFY, HSFY2ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-57163