Learn More
Aviva Systems Biology Corp HSFY2 Antibody - C-terminal region (ARP37380_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37380_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HSFY2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Hspy2l, 4933413G11RikApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56357