Learn More
Medchemexpress LLC HSGN-94 | 2570797-85-0 | 508.51 | 50 MG
SDP
Catalog No. 5000361493
Encompass_Preferred
HSGN-94 is a potent antimicrobial agent with lipoteichoic acid (LTA) biosynthesis inhibition. HSGN-94 inhibits drug-resistant Gram-positive bacteria with MIC values of 0.25-2 μg/mL. HSGN-94 inhibits biofilm formation of MRSA and Vancomycin-resistant Enterococci. HSGN-94 also inhibits pro-inflammatory cytokines, exhibits in vivo efficacy in an MRSA murine wound infection model.
  • Potent antimicrobial agent with LTA biosynthesis inhibition.
  • Inhibits drug-resistant Gram-positive bacteria with MIC values of 0.25-2 μg/mL.
  • Inhibits biofilm formation in MRSA and Vancomycin-resistant Enterococci.
  • Inhibits pro-inflammatory cytokines.
  • Directly binds to PgsA.
  • Downregulates the expression level of PgsA, an essential protein for phospholipid synthesis.
Catalog No. 50-003-61493 Supplier Medchemexpress LLC Supplier No. HY14681150MG
HSGN-94 is a potent antimicrobial agent with lipoteichoic acid (LTA) biosynthesis inhibition. HSGN-94 inhibits drug-resistant Gram-positive bacteria with MIC values of 0.25-2 μg/mL. HSGN-94 inhibits biofilm formation of MRSA and Vancomycin-resistant Enterococci. HSGN-94 also inhibits pro-inflammatory cytokines, exhibits in vivo efficacy in an MRSA murine wound infection model.
  • Potent antimicrobial agent with LTA biosynthesis inhibition.
  • Inhibits drug-resistant Gram-positive bacteria with MIC values of 0.25-2 μg/mL.
  • Inhibits biofilm formation in MRSA and Vancomycin-resistant Enterococci.
  • Inhibits pro-inflammatory cytokines.
  • Directly binds to PgsA.
  • Downregulates the expression level of PgsA, an essential protein for phospholipid synthesis.