Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HSGN-94 | 2570797-85-0 | 98.8% | C23H23F3N4O4S | 10MG
SDP

Catalog No. 5000259176
Encompass_Preferred

HSGN-94 is a small-molecule antimicrobial research compound that inhibits lipoteichoic acid (LTA) biosynthesis and demonstrates activity against drug-resistant Gram-positive bacteria. It is provided as a powder or as a DMSO solution for in vitro antimicrobial and anti-infection pathway studies.

  • Inhibits lipoteichoic acid biosynthesis.
  • Active against drug-resistant Gram-positive bacteria.
  • Supplied as powder and as DMSO solution for flexibility in assays.
  • High purity suitable for research applications.
  • Soluble in DMSO; ultrasonic agitation may aid dissolution.
  • Recommended storage at low temperature to preserve stability.

Catalog No. 50-002-59176 Supplier Medchemexpress LLC Supplier No. HY14681110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HSGN-94 is a small-molecule antimicrobial research compound that inhibits lipoteichoic acid (LTA) biosynthesis and demonstrates activity against drug-resistant Gram-positive bacteria. It is provided as a powder or as a DMSO solution for in vitro antimicrobial and anti-infection pathway studies.

  • Inhibits lipoteichoic acid biosynthesis.
  • Active against drug-resistant Gram-positive bacteria.
  • Supplied as powder and as DMSO solution for flexibility in assays.
  • High purity suitable for research applications.
  • Soluble in DMSO; ultrasonic agitation may aid dissolution.
  • Recommended storage at low temperature to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.