Learn More
Aviva Systems Biology Corp HSH2D Antibody - C-terminal region (ARP60696_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60696_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HSH2D. It was validated on Western Blot. . Alternate names: ALX, FLJ14886, HSH2ApplicationWB. Species reactivity: Human
Numéro de catalogue. 50-287-129
Ajouter au panier
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.