Learn More
Medchemexpress LLC HY-101509 5mg Medchemexpress, HSL-IN-1 CAS:2095156-13-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015095MG
Encompass_Preferred
Medchemexpress, HY-101509 5mg HSL-IN-1 CAS:2095156-13-9 HSL-IN-1 (compound 24b) is a potent and orally active hormone sensitive lipase (HSL) inhibitor (IC50=2 nM) with a significantly reduced reactive metabolite liability. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9496