Learn More
Medchemexpress LLC HY-102056 5mg Medchemexpress, HSL-IN-2 CAS:654059-21-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020565MG
Encompass_Preferred
Medchemexpress, HY-102056 5mg HSL-IN-2 CAS:654059-21-9 HSL-IN-2 is a potent and selective inhibitor of Hormone Sensitive Lipase (HSL), with an IC50 of 0.023 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7974