Learn More
TARGETMOL CHEMICALS INC HSL-IN-3 5MG
SDP
Catalog No. 502581521
Encompass_Preferred
Also available in 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HSL-IN-3 (HSL inhibitor 3) is a boronic acid derivative that is an inhibitor of hormone-sensitive lipase(HSL). purity: 95%
Catalog No. 50-258-1521 Supplier TARGETMOL CHEMICALS INC Supplier No. T115795MG
Also available in 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HSL-IN-3 (HSL inhibitor 3) is a boronic acid derivative that is an inhibitor of hormone-sensitive lipase(HSL). purity: 95%