Learn More
Enterprise Technology Solutions HSM Shredder, Cross-cut, Anti-Jam, 17-Sht Cap, White
SDP
Catalog No. 5000740533
Encompass_Preferred
The epitome of function meets affordability, the shredstar X17 provides quiet and powerful data security. Simple-to-use features come in a convenient design. It is ideal for personal/home office use. Shredder powers through 17 sheets of paper, creating 407 cross-cut/confetti-cut particles per page and dropping them into a 6.9 gallon bin. Monitor the fill level through the inspection window. The separate CD cutting unit has its own waste container so you don't have to worry about it mixing with the paper. Shredder is also equipped with anti-jam technology, LED indicator to show operating status and light barrier for automatic start/stop. Quiet operation minimizes noise in your office area.
Catalog No. 50-007-40533 Supplier Enterprise Technology Solutions Supplier No. HSM1030133
The epitome of function meets affordability, the shredstar X17 provides quiet and powerful data security. Simple-to-use features come in a convenient design. It is ideal for personal/home office use. Shredder powers through 17 sheets of paper, creating 407 cross-cut/confetti-cut particles per page and dropping them into a 6.9 gallon bin. Monitor the fill level through the inspection window. The separate CD cutting unit has its own waste container so you don't have to worry about it mixing with the paper. Shredder is also equipped with anti-jam technology, LED indicator to show operating status and light barrier for automatic start/stop. Quiet operation minimizes noise in your office area.