Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC hSMG-1 inhibitor 11e | 1402452-10-1 | 99.9% | 517.60 | C26H27N7O3S | 100MG
SDP

Catalog No. 5000238267
Encompass_Preferred

hSMG-1 inhibitor 11e is a research-grade, potent and selective inhibitor of the human SMG-1 kinase. It exhibits sub-nanomolar potency (hSMG-1 IC50 <0.05 nM) and pronounced selectivity versus related kinases, and is supplied as a solid or as a 10 mM solution in DMSO for biochemical and cellular studies.

  • Potent hSMG-1 inhibition (IC50 <0.05 nM).
  • High selectivity (>900-fold) over mTOR, with reported IC50s for mTOR 45 nM, PI3Kα 61 nM, PI3Kγ 92 nM.
  • Molecular formula C26H27N7O3S; molecular weight 517.60.
  • Purity 99.9% (manufacturer listed 99.94%, rounded to tenths).
  • Available as solid (including 100 mg) and 10 mM solution in DMSO.
  • Suitable for biochemical kinase assays and cellular pathway studies.

Catalog No. 50-002-38267 Supplier Medchemexpress LLC Supplier No. HY124760100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

hSMG-1 inhibitor 11e is a research-grade, potent and selective inhibitor of the human SMG-1 kinase. It exhibits sub-nanomolar potency (hSMG-1 IC50 <0.05 nM) and pronounced selectivity versus related kinases, and is supplied as a solid or as a 10 mM solution in DMSO for biochemical and cellular studies.

  • Potent hSMG-1 inhibition (IC50 <0.05 nM).
  • High selectivity (>900-fold) over mTOR, with reported IC50s for mTOR 45 nM, PI3Kα 61 nM, PI3Kγ 92 nM.
  • Molecular formula C26H27N7O3S; molecular weight 517.60.
  • Purity 99.9% (manufacturer listed 99.94%, rounded to tenths).
  • Available as solid (including 100 mg) and 10 mM solution in DMSO.
  • Suitable for biochemical kinase assays and cellular pathway studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.