Learn More
Medchemexpress LLC hSMG-1 inhibitor 11e | 1402452-10-1 | 99.9% | 517.60 g/mol | C26H27N7O3S | 10MG
SDP
Catalog No. 5000239089
Encompass_Preferred
hSMG-1 inhibitor 11e is a potent, selective small-molecule inhibitor of human SMG-1 kinase with sub-nanomolar activity. It demonstrates strong selectivity versus related PI3K-like kinases and CDKs and is supplied as a high-purity research-grade compound with supporting analytical documentation for biochemical and cellular studies.
  • Sub-nanomolar potency against hSMG-1 (IC50 <0.05 nM).
  • Selective versus mTOR and PI3K isoforms with reported >900-fold selectivity.
  • High reported purity suitable for research applications.
  • Provided with supporting analytical files, including COA, LCMS, and HNMR.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-39089 Supplier Medchemexpress LLC Supplier No. HY12476010MG
hSMG-1 inhibitor 11e is a potent, selective small-molecule inhibitor of human SMG-1 kinase with sub-nanomolar activity. It demonstrates strong selectivity versus related PI3K-like kinases and CDKs and is supplied as a high-purity research-grade compound with supporting analytical documentation for biochemical and cellular studies.
  • Sub-nanomolar potency against hSMG-1 (IC50 <0.05 nM).
  • Selective versus mTOR and PI3K isoforms with reported >900-fold selectivity.
  • High reported purity suitable for research applications.
  • Provided with supporting analytical files, including COA, LCMS, and HNMR.
  • Intended for research use only; not for human or clinical use.