Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HSMG-1 inhibitor 11e | 1402452-10-1 | 99.9% | 517.60 g/mol | C26H27N7O3S | 25MG
SDP

Catalog No. 5000238740
Encompass_Preferred

hSMG-1 inhibitor 11e is a potent, selective small-molecule inhibitor of human SMG-1 kinase with sub-nanomolar activity (IC50 <0.05 nM). It demonstrates strong selectivity versus related kinases, is supplied as a solid (white to light yellow), and is intended for research use. The compound is soluble in DMSO and has defined storage recommendations for both solid and solution forms.

  • Sub-nanomolar potency (hSMG-1 IC50 <0.05 nM).
  • High selectivity over several off-target kinases (mTOR, PI3Kα/γ, CDK1/2).
  • High purity (99.94%).
  • Good DMSO solubility (50 mg/mL) for in vitro applications.
  • Recommended storage: solid at 4°C; in solvent at -80°C for long-term stability.
  • White to light yellow solid appearance.
  • Molecular weight 517.60 g/mol; formula C26H27N7O3S.

Catalog No. 50-002-38740 Supplier Medchemexpress LLC Supplier No. HY12476025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

hSMG-1 inhibitor 11e is a potent, selective small-molecule inhibitor of human SMG-1 kinase with sub-nanomolar activity (IC50 <0.05 nM). It demonstrates strong selectivity versus related kinases, is supplied as a solid (white to light yellow), and is intended for research use. The compound is soluble in DMSO and has defined storage recommendations for both solid and solution forms.

  • Sub-nanomolar potency (hSMG-1 IC50 <0.05 nM).
  • High selectivity over several off-target kinases (mTOR, PI3Kα/γ, CDK1/2).
  • High purity (99.94%).
  • Good DMSO solubility (50 mg/mL) for in vitro applications.
  • Recommended storage: solid at 4°C; in solvent at -80°C for long-term stability.
  • White to light yellow solid appearance.
  • Molecular weight 517.60 g/mol; formula C26H27N7O3S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.