Learn More
TARGETMOL CHEMICALS INC HSMG-1 INHIBITOR 11E 5MG
SDP
Catalog No. 502580218
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hSMG-1 inhibitor 11e is an effective and selective inhibitor of hSMG-1 (IC50 <0.05 nM) and can be used in studies about cancer treatment. purity: 98%
Catalog No. 50-258-0218 Supplier TARGETMOL CHEMICALS INC Supplier No. T355295MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. hSMG-1 inhibitor 11e is an effective and selective inhibitor of hSMG-1 (IC50 <0.05 nM) and can be used in studies about cancer treatment. purity: 98%