Learn More
New England Biolabs, Inc. hSMUG1 – 500 units
SDP
Catalog No. 504378
Encompass
hSMUG1 excises deoxyuracil and deoxyuracil-derivatives leaving an oxidized group at C5, like 5-hydroxyuracil, 5-hydroxymethyluracil and 5-formyluracil.
Catalog No. 50-437-8 Supplier New England Biolabs, Inc. Supplier No. M0336S
hSMUG1 excises deoxyuracil and deoxyuracil-derivatives leaving an oxidized group at C5, like 5-hydroxyuracil, 5-hydroxymethyluracil and 5-formyluracil.