Learn More
BioVendor, LLC Recombinant protein 0.1 mg, 1/ea
SDP
Supplier:  BioVendor, LLC RD172263100
Encompass
Recombinant protein Description Total 111 AA. MW: 12.0 kDa (calculated). UniProtKB acc.no. P61604. N-Terminal His-tag (10 extra AA)Amino Acid SequenceMKHHHHHHASAGQAFRKFLPLFDRVLVERSAAETVTKGGIMLPEKSQGKVLQATVVAVGSGSKGKGGEIQPVSVKVGDKVLLPEYGGTKVVLDDKDYFLFRDGDILGKYVDSourceE. coliPurity: >95% Endotoxin< 0.1 EU/?gFormulationFiltered (0,4 ?m) and lyophilized in 0.5 mg/mL in 0.05 M phosphate buffer, 0.075 M NaCl, pH 7.4ReconstitutionAdd deionized water to prepare a working stock solution of approximately 0.5 mg/mL and let the lyophilized pellet dissolve completely. Filter sterilize your culture media/working solutions containing this non-sterile product before using in cell culture.ApplicationsWestern blotting, ELISAQuality Control TestBCA to determine quantity of the protein.SDS PAGE to determine purity of the protein.LAL to determine quantity of endotoxin.
Catalog No. 50-384-2