Learn More
Enzo Life Sciences HSP104 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210140R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 894-908 (D894DDNEDSMEIDDDLD908) of C-terminal S. cerevisiae HSP104 (heat shock protein 104). Species reactivity: Yeast. Applications: WB. Formulation: Liquid. Contains 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8077