Learn More
Enzo Life Sciences HSP 110 polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA1101E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of hamster HSP110. The sequence is completely conserved in human and mouse. UniProt: Q60446. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Yeast, Sheep, Monkey, Hamster, Bovine. Applications: WB. Recommended dilutions: Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-418