Learn More
Enzo Life Sciences HSP20 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA796F
Encompass_Preferred
Host: Rabbit. Immunogen: Rat skeletal muscle Hsp20. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Fish, Dog, Bovine. Applications: WB. Purity: Peptide affinity purified. Formulation: Liquid. In 0.1M phosphate buffer, pH 7.0, containing 3% BSA and 0.09% sodium azide.
Catalog No. 50-200-7470