Learn More
Enzo Life Sciences HSP25 (mouse), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP510D
Encompass_Preferred
Alternative name: HspB1, Heat shock protein 25. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~28kDa. Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7604