Learn More
Enzo Life Sciences HSP25 polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISPA801J
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant mouse Hsp25. UniProt ID: P14602. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Porcine, Hamster, Guinea pig, Gerbil, Dog, Bovine. Applications: ICC, IF, IP, WB Electron microscopy. Recommended Dilutions: Immunoprecipitation (1:100) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7478