Learn More
Enzo Life Sciences HSP 25 polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA801D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant mouse Hsp25. UniProt: P14602. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Porcine, Hamster, Guinea pig, Gerbil, Dog, Bovine. Applications: ICC, IF, IP, WB, Electron microscopy. Recommended dilutions: Immunoprecipitation (1:100). Western Blot (1:1,000, colorimetric). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-530