Learn More
Enzo Life Sciences HSP 27 monoclonal antibody (G3.1) (biotin conjugate) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA800BD
Encompass_Preferred
Clone: G3.1. Host: Mouse. Isotype: IgG1. Immunogen: Native human HSP27. UniProt: P04792. Source: Purified from mouse ascites. Species Reactivity: Human, Mouse, Rat, Monkey, Fish, Bovine. Applications: WB. Recommended dilutions: Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-520