Learn More
Enzo Life Sciences IMMUNOSET HSP27 high sensitivity (human), ELISA development set (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI960076
Encompass_Preferred
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available. The IMMUNOSET HSP27 high sensitivity (human), EIA development set contains sufficient reagents for the development of 5 x 96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules. Alternative name: Heat shock protein 27. Content: Capture antibody, Standard, Detection antibody, SA-HRP. Long Term Storage: +4°C. Creossreactivity: Specific for HSP27. Does not cross-react with HSP25, Cpn10, HO-1, HSP40, or αB-crystallin. Assay Time: Plate Coating - Overnight + 1 hour; Assay - 2 hours 45 minutes. Applications: ELISA, Colorimetric detection .
Catalog No. 50-200-7128