Learn More
Enzo Life Sciences HSP27 (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS500
Encompass_Preferred
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available. The HSP27 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 450 nm. Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis. Highly sensitive quantitative determination of Hsp27 independent of phosphorylation state Alternative name: Heat shock protein 27. Sensitivity: 0.39 ng/ml (range 0.39 - 25 ng/ml). Use/Stability: Store all components at 4°C, except standard at -20°C. Assay Time: <3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-200-7215