Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HSP27 inhibitor J2 | 2133499-85-9 | 99.7% | 264.30 g/mol | C13H12O4S | 25MG
SDP

Catalog No. 5000235127
Encompass_Preferred

HSP27 inhibitor J2 is a small-molecule research reagent that inhibits heat shock protein 27 (HSP27) by inducing abnormal dimer formation and disrupting HSP27 polymer function. It is supplied as a purified solid for biochemical and cellular studies with documented solubility and storage recommendations.

  • High purity (99.7%).
  • Molecular weight 264.30 g/mol.
  • Chemical formula C13H12O4S.
  • Soluble in DMSO at 20 mg/mL (75.67 mM); hygroscopic DMSO may affect solubility.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 2 years) or -20°C (up to 1 year).
  • Available in multiple pack sizes including 25 mg.

Catalog No. 50-002-35127 Supplier Medchemexpress LLC Supplier No. HY12465325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HSP27 inhibitor J2 is a small-molecule research reagent that inhibits heat shock protein 27 (HSP27) by inducing abnormal dimer formation and disrupting HSP27 polymer function. It is supplied as a purified solid for biochemical and cellular studies with documented solubility and storage recommendations.

  • High purity (99.7%).
  • Molecular weight 264.30 g/mol.
  • Chemical formula C13H12O4S.
  • Soluble in DMSO at 20 mg/mL (75.67 mM); hygroscopic DMSO may affect solubility.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 2 years) or -20°C (up to 1 year).
  • Available in multiple pack sizes including 25 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.