Learn More
Enzo Life Sciences HSP27 monoclonal antibody (G3.1) (FITC conjugate) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA800FID
Encompass_Preferred
Clone: G3.1. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp27. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Monkey, Fish, Bovine. Applications: Flow Cytometry. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles. Protect from light.
Catalog No. 50-200-7473