Learn More
Enzo Life Sciences HSP27 monoclonal antibody (G3.1) (PE conjugate) (100µg)
SDP
Supplier:  Enzo Life Sciences ADISPA800PEE
Encompass_Preferred
Clone: G3.1. Host: Mouse. Isotype: IgG1. Immunogen: Native human Hsp27. UniProt ID: P04792. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Monkey, Fish, Bovine. Applications: Flow Cytometry. Recommended Dilutions: Flow Cytometry (10µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide. Handling: Avoid vigorous centrifugation and vortexing. Do not freeze. Protect from light.
Catalog No. 50-200-7475