Learn More
Enzo Life Sciences HSP27 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA803D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Hsp27. Source: Purified from rabbit serum. Species reactivity: Human Porcine, Monkey. Applications: ELISA, IHC, IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7481