Learn More
Enzo Life Sciences HSP27 (phospho) (human), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP716F
Encompass_Preferred
Alternative name: HspB1, Heat shock protein 27. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~27kDa. Formulation: Liquid. In 60mM Tris-HCl, pH 7.5, containing 5mM sodium chloride, 1mM EGTA, 0.5mM EDTA, 15mM DTT, 0.1mM ATP, and 15mM magnesium chloride. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7624