Learn More
Enzo Life Sciences HSP27 (pSer15) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA525F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human HSP27 phosphorylated at Ser15. UniProt ID: P04792. Source: Purified from rabbit serum. Species reactivity: Human, Monkey, Guinea pig, Dog, Bovine. Applications: IF, IHC, WB. Recommended Dilutions: Immunohistochemistry (1:50) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7441