Learn More
Enzo Life Sciences HSP27 (pSer15) polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA525D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human HSP27 phosphorylated at Ser15. Source: Purified from rabbit serum. Species reactivity: Human, Monkey, Guinea pig, Dog, Bovine. Applications: IF, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7440