Learn More
Enzo Life Sciences HSP27 (pSer78) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA523F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human HSP27 phosphorylated at Ser78. Source: Purified from rabbit serum. Species reactivity: Human, Rat, Monkey, Guinea pig, Bovine. Applications: IHC, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7439