Learn More
Enzo Life Sciences HSP27 (pSer82) monoclonal antibody (5B9) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804588C100
Encompass_Preferred
Clone: 5B9. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 79-85 (R79QLpSSGV85) of human HSP27 (heat shock protein 27) phosphorylated at Ser82 conjugated to KLH. Species reactivity: Human, Mouse, Rat, Dog. Specificity: Recognizes HSP27 phosphorylated at Ser82. Applications: IHC, WB. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose.
Catalog No. 50-200-9952