Learn More
Cayman Chemical Hsp27HspB1 Monclonal ntI 100ug
SDP
Catalog No. 502936066
Encompass_Preferred
A depsidone with antibacterial activity inhibits the growth of MRSA (MIC 2 Ug/ml) has cytotoxic activity in MOLT-3 cells (IC50 35.7 UM)
Catalog No. 50-293-6066 Supplier Cayman Chemical Supplier No. 25692100 UG
A depsidone with antibacterial activity inhibits the growth of MRSA (MIC 2 Ug/ml) has cytotoxic activity in MOLT-3 cells (IC50 35.7 UM)