Learn More
Enzo Life Sciences HSP40 Hdj1 monoclonal antibody (2E1) (100µg)
SDP
Supplier:  Enzo Life Sciences ADISPA450E
Encompass_Preferred
Clone: 2E1. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human Hsp40 (Hdj1). Species reactivity: Human, Mouse, Rat. Applications: IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7435