Learn More
Enzo Life Sciences HSP40 Hdj1 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA400D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human Hsp40. UniProt ID: P25685. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Scallop, Rabbit, Porcine, Mussel, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine, Beluga. Applications: ICC, IF, IHC (PS), IP, WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7429