Learn More
Enzo Life Sciences HSP40 Hdj2 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA405050
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human HSP40/Hdj2. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Hamster. Specificity: Has high specificity for the constitutively expressed Hdj2 compared to Hdj1 using recombinant controls. Applications: WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7431