Learn More
Enzo Life Sciences HSP40 (human), (recombinant) (His-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX201274C100
Encompass_Preferred
Alternative name: Heat shock protein 40, Hdj1, DnaJ protein homolog 1. Concentration: 1mg/ml. Endotoxin: <0.1EU/μg protein (determined by LAL method). Purity: ≥95% (SDS-PAGE). MW: ~40.2kDa. Formulation: Liquid. In 20mM TRIS, pH 8.0, containing 2mM DDT, 0.2mM PMSF and 10% glycerol. Source: Produced in E.coli and fused to a His-tag. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7963