Learn More
Enzo Life Sciences HSP 40 Hdj1 (human), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP400F
Encompass_Preferred
Alternative name: Heat shock protein 40. DnaJ protein homolog 1. MW: ~40kDa. Purity: ≥80% (SDS-PAGE). Formulation: Liquid. In Dulbecco’s PBS. Source: Produced in E. coli. UniProt: P25685. Application: WB, in vitro Assay, EMSA, Antigen microarray. Long Term Storage: -80°C
Catalog No. 50-667-714