Learn More
Enzo Life Sciences HSP 47 monoclonal antibody (M16.10A1) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPA470F
Encompass_Preferred
Clone: M16.10A1. Host: Mouse. Isotype: IgG2bλ. Immunogen: Native rat HSP47. UniProt: P50454. Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Dog, Chicken, Bovine. Applications: IHC, WB. Recommended dilutions: Immunohistochemistry (1:50). Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-464