Learn More
Enzo Life Sciences HSP 47 (human), (recombinant) (200 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP535F
Encompass_Preferred
Alternative name: SerpinH1, Colligin, Gp46, Heat shock protein 47. Collagen binding protein. MW: ~47kDa. Endotoxin content: < 50 EU/mg determined by Limulus Amebocyte Lysate (LAL) gel clot assay. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 30mM TRIS, pH 8.0, containing 100mM sodium chloride, 1mM DTT, and 0.4M arginine. Source: Produced in E. coli. UniProt: P50454. Application: WB, Antigen microarray. Long Term Storage: -80°C
Catalog No. 50-667-722