Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3329MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM253. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: HeLa or HepG2 cells. Synovial biopsies from patients with juvenile chronic arthritis. Synovial lining layer is strongly positive for hsp60. Breast carcinoma. Cellular Staining: Mitochondria in cytoplasm. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human HSP60 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 60kDa.Chromosome: 2q33.1. Gene Symbol: HSPD1. Gene ID: 3329. Ensemble: ENSG00000144381. Uniprot: NP_955472.
Catalog No. 50-188-4784