Learn More
Enzo Life Sciences Anti-HSP60 IgG/A/M (human), ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADIEKS650
Encompass_Preferred
Most comprehensive portfolio of heat shock protein (chaperone) products commercially available. Sensitivity: 2.88 ng/ml (range 7.81 - 250 ng/ml). Assay Time: <4 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Standard, Sample diluent 1, Wash buffer concentrate, HRP Conjugate, TMB Substrate, Stop solution 2
Catalog No. 50-659-132