Learn More
Enzo Life Sciences HSP60 (bacterial) monoclonal antibody (A57-B9) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804072R100
Encompass_Preferred
Clone: A57-B9. Isotype: IgG1. Immunogen: Recombinant Chlamydia trachomatis HSP60 (heat shock protein 60). Species reactivity: Chlamydia. Specificity: Recognizes all three genera of Chlamydia HSP60. Epitope mapping studies suggest binding of aa 517-522. Applications: ICC, IP, WB. Formulation: Liquid. Ascites diluted in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9769