Learn More
Enzo Life Sciences HSP60 (bacterial) monoclonal antibody (A57-E4) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX804071R100
Encompass_Preferred
Clone: A57-E4. Isotype: IgG1. Immunogen: Recombinant Chlamydia trachomatis HSP60 (heat shock protein 60). Species reactivity: Salmonella typhimurium, Neisseria gonorrhoeae, E. coli, Coxiella burnetti, Chlamydia. Specificity: Recognizes bacterial HSP60 from Chlamydia, E. coli (GroEL), Salmonella typhimurium, Neisseria gonorrhoeae, and Coxiella burnetti. Applications: WB. Formulation: Liquid. Ascites diluted in PBS containing 0.05% sodium azide.
Catalog No. 50-200-9768