Learn More
Enzo Life Sciences HSP60 (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP540050
Encompass_Preferred
Alternative name: Chaperonin 60, CPN60, HspD1, Heat shock protein 60. Purity: ≥90% (SDS-PAGE; Western blot). MW: ~60kDa. Formulation: Liquid. In Dulbecco's PBS. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7606